Learn More
Electron Microscopy Sciences Wet or Dry Silicone Carbide Disc 8", 800 Grit, PSA backing
SDP
Catalog No. 50997159
Encompass_Preferred
Pregrinding Wet/Dry Abrasive Papers (Discs) with premium Resin Bonding System for longer wear. Special adhesive eliminates need to transfer to plates. Uniform mineral size. Pull tabs for easy use.
Particles: 800 Grit
Backing: PSA
Catalog No. 50-997-159 Supplier Electron Microscopy Sciences Supplier No. 5025509
Pregrinding Wet/Dry Abrasive Papers (Discs) with premium Resin Bonding System for longer wear. Special adhesive eliminates need to transfer to plates. Uniform mineral size. Pull tabs for easy use.
Particles: 800 Grit
Backing: PSA