Learn More
Electron Microscopy Sciences Silicone Carbide Dressing Stick
SDP
Catalog No. 50997381
Encompass_Preferred
Made from silicon carbide. Use for cutting blade dressing.
Catalog No. 50-997-381 Supplier Electron Microscopy Sciences Supplier No. 5037901
Made from silicon carbide. Use for cutting blade dressing.