Learn More
Electron Microscopy Sciences Silicone Carbide Powder, 42 µm particles 1 LB
SDP
Catalog No. 50997180
Encompass_Preferred
Dark gray 98% SiC. Hardness, Moh's scale 9.0; Knoop Scale 2500. Crystallography: hexagonal–alpha and beta phase.
Size: 42 µm
Catalog No. 50-997-180 Supplier Electron Microscopy Sciences Supplier No. 5026042
Dark gray 98% SiC. Hardness, Moh's scale 9.0; Knoop Scale 2500. Crystallography: hexagonal–alpha and beta phase.
Size: 42 µm