Learn More
Electron Microscopy Sciences Silicone Carbide Powder, 50 µm particles 1 LB
SDP
Catalog No. 50997181
Encompass_Preferred
Dark gray 98% SiC. Hardness, Moh's scale 9.0; Knoop Scale 2500. Crystallography: hexagonal–alpha and beta phase.
Size: 50 µm
Catalog No. 50-997-181 Supplier Electron Microscopy Sciences Supplier No. 5026050
Dark gray 98% SiC. Hardness, Moh's scale 9.0; Knoop Scale 2500. Crystallography: hexagonal–alpha and beta phase.
Size: 50 µm