Learn More
Bellco Glass, Inc. Silicone Closure,18-25mm
SDP
Catalog No. 501945239
Encompass_Preferred
Sealed in silicone sponge closure is ideal for aerobic cultures. Flanged to fit fit snugly over outside of the neck preventing passage of aerosols. Silicone sponge closures can withstand dry heat sterilization up to 180 degrees Celsius. Use mainly for flasks, to prevent any trouble due to tumbling of the flask.Specifications:Cap TypeAir Permeation: 200-300 (Cc/Min)For Use With: 18-25MM CULTURE TUBES / 25-50ML ERLENMEYER FLASKS / 25-125ML DELONG FLASKSCase Quantity: 10
Catalog No. 50-194-5239 Supplier Bellco Glass, Inc. Supplier No. 200400003
Sealed in silicone sponge closure is ideal for aerobic cultures. Flanged to fit fit snugly over outside of the neck preventing passage of aerosols. Silicone sponge closures can withstand dry heat sterilization up to 180 degrees Celsius. Use mainly for flasks, to prevent any trouble due to tumbling of the flask.Specifications:Cap TypeAir Permeation: 200-300 (Cc/Min)For Use With: 18-25MM CULTURE TUBES / 25-50ML ERLENMEYER FLASKS / 25-125ML DELONG FLASKSCase Quantity: 10