Learn More
Chemglass Life Sciences Closure, Silicone Sponge, 20mm
SDP
Catalog No. 501215153
Encompass_Preferred
Closure, Silicone Sponge, 20mm. The silicone sponge closure is manufactured with a sealed in silicone sponge filter. Flange fits snugly over the necks of vessels permitting the exchange of gases, while reducing evaporation and preventing the passing of aerosols. Ideal for use with aerobic cultures. Reusable, autoclavable and will withstand dry heat sterilization up to 180. Closure will withstand both dry heat sterilization and repeated autoclaving.CLS-1490-020 fits: 18-25mm Culture Tubes, 25-50mL Erlenmeyer Flasks and 25-125mL Delong Flasks
Catalog No. 50-121-5153 Supplier Chemglass Life Sciences Supplier No. CLS1490020
Closure, Silicone Sponge, 20mm. The silicone sponge closure is manufactured with a sealed in silicone sponge filter. Flange fits snugly over the necks of vessels permitting the exchange of gases, while reducing evaporation and preventing the passing of aerosols. Ideal for use with aerobic cultures. Reusable, autoclavable and will withstand dry heat sterilization up to 180. Closure will withstand both dry heat sterilization and repeated autoclaving.CLS-1490-020 fits: 18-25mm Culture Tubes, 25-50mL Erlenmeyer Flasks and 25-125mL Delong Flasks