Learn More
Chemglass Life Sciences Closure, Silicone Sponge, 28mm
SDP
Catalog No. 501215154
Encompass_Preferred
Closure, Silicone Sponge, 28mm. The silicone sponge closure is manufactured with a sealed in silicone sponge filter. Flange fits snugly over the necks of vessels permitting the exchange of gases, while reducing evaporation and preventing the passing of aerosols. Ideal for use with aerobic cultures. Reusable, autoclavable and will withstand dry heat sterilization up to 180°. Closure will withstand both dry heat sterilization and repeated autoclaving.CLS-1490-028 fits: 125 and 250mL Erlenmeyer Flasks
Catalog No. 50-121-5154 Supplier Chemglass Life Sciences Supplier No. CLS1490028
Closure, Silicone Sponge, 28mm. The silicone sponge closure is manufactured with a sealed in silicone sponge filter. Flange fits snugly over the necks of vessels permitting the exchange of gases, while reducing evaporation and preventing the passing of aerosols. Ideal for use with aerobic cultures. Reusable, autoclavable and will withstand dry heat sterilization up to 180°. Closure will withstand both dry heat sterilization and repeated autoclaving.CLS-1490-028 fits: 125 and 250mL Erlenmeyer Flasks