Learn More
Chemglass Life Sciences Closure, Silicone Sponge, 38mm
SDP
Catalog No. 501215155
Encompass_Preferred
Closure, Silicone Sponge, 38mm. The silicone sponge closure is manufactured with a sealed in silicone sponge filter. Flange fits snugly over the necks of vessels permitting the exchange of gases, while reducing evaporation and preventing the passing of aerosols. Ideal for use with aerobic cultures. Reusable, autoclavable and will withstand dry heat sterilization up to 180. Closure will withstand both dry heat sterilization and repeated autoclaving.CLS-1490-038 fits: 38mm Delong Style Flask or Tube Necks, 38mm Culture Tubes, 500 and 1000mL Standard Erlenmeyer Flasks and GL-45 Screw Threads
Catalog No. 50-121-5155 Supplier Chemglass Life Sciences Supplier No. CLS1490038
Add to Cart
Add to Cart
Closure, Silicone Sponge, 38mm. The silicone sponge closure is manufactured with a sealed in silicone sponge filter. Flange fits snugly over the necks of vessels permitting the exchange of gases, while reducing evaporation and preventing the passing of aerosols. Ideal for use with aerobic cultures. Reusable, autoclavable and will withstand dry heat sterilization up to 180. Closure will withstand both dry heat sterilization and repeated autoclaving.CLS-1490-038 fits: 38mm Delong Style Flask or Tube Necks, 38mm Culture Tubes, 500 and 1000mL Standard Erlenmeyer Flasks and GL-45 Screw Threads