Learn More
Bellco Glass, Inc. Silicone Closure,38mm
SDP
Catalog No. 501945241
Encompass_Preferred
Sealed in silicone sponge closure is ideal for aerobic cultures. Flanged to fit fit snugly over outside of the neck preventing passage of aerosols. Silicone sponge closures can withstand dry heat sterilization up to 180 degrees Celsius. Use mainly for flasks, to prevent any trouble due to tumbling of the flask.Specifications:Cap TypeAir Permeation: 200-300 (Cc/Min)For Use With: 38MM CULTURE TUBES / 500-1000ML ERLENMEYER FLASKS / 250-6000ML DELONG FLASKSCase Quantity: 10
Catalog No. 50-194-5241 Supplier Bellco Glass, Inc. Supplier No. 200400005
Sealed in silicone sponge closure is ideal for aerobic cultures. Flanged to fit fit snugly over outside of the neck preventing passage of aerosols. Silicone sponge closures can withstand dry heat sterilization up to 180 degrees Celsius. Use mainly for flasks, to prevent any trouble due to tumbling of the flask.Specifications:Cap TypeAir Permeation: 200-300 (Cc/Min)For Use With: 38MM CULTURE TUBES / 500-1000ML ERLENMEYER FLASKS / 250-6000ML DELONG FLASKSCase Quantity: 10