Learn More
Perkin Elmer US LLC Silicone Crystal with Holder - Spare
Catalog No. 502117266
Encompass_Preferred
The Micro ATR Objective Upgrade Kit upgrades Spotlight 400, Spotlight 200i, Spotlight 100i, and microscope systems without an ATR capibility with a micro ATR facility that covers the range 5,500 600 cm-1.
Catalog No. 50-211-7266 Supplier Perkin Elmer US LLC Supplier No. L1860269
The Micro ATR Objective Upgrade Kit upgrades Spotlight 400, Spotlight 200i, Spotlight 100i, and microscope systems without an ATR capibility with a micro ATR facility that covers the range 5,500 600 cm-1.