Learn More
Crescent Chemical Co Inc SILICONE DC 200 FLUID; 100 CST
SDP
Catalog No. 50248146
Encompass_Preferred
Silicone DC 200 fluid; 100 cst pract. Dimethyl siloxane polymer (methyl silicones).In terms of SI-units: 1 cst = 10-6m2s-1Density (25 °C) 0.962 - 0.968 Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35135.01
Catalog No. 50-248-146 Supplier Crescent Chemical Co Inc Supplier No. SE35135.01
Silicone DC 200 fluid; 100 cst pract. Dimethyl siloxane polymer (methyl silicones).In terms of SI-units: 1 cst = 10-6m2s-1Density (25 °C) 0.962 - 0.968 Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35135.01