Learn More
Crescent Chemical Co Inc SILICONE DC 710 FLUID; 500 CST
SDP
Catalog No. 50248150
Encompass_Preferred
Silicone DC 710 fluid; 500 cst pract. (Phenylmethyldimethylsiloxane)In terms of SI-units: 1 cst = 10 -6m2s-1(50 % Methyl-50 % phenylsilicone). For GC, tmax. 225 °. Extremely stable at high temperatures.Density (25 °C) 1.10 - 1.11 Size - 1KG Storage Conditions - +15 °C
Catalog No. 50-248-150 Supplier Crescent Chemical Co Inc Supplier No. SE35149.04
Silicone DC 710 fluid; 500 cst pract. (Phenylmethyldimethylsiloxane)In terms of SI-units: 1 cst = 10 -6m2s-1(50 % Methyl-50 % phenylsilicone). For GC, tmax. 225 °. Extremely stable at high temperatures.Density (25 °C) 1.10 - 1.11 Size - 1KG Storage Conditions - +15 °C