Learn More
World Precision Instrument Silicone Dissecting Pad Kit
SDP
Catalog No. NC9666100
Encompass
Make your own silicone dissecting pads easily and quickly. Mix the 2-part silicone right in the plastic petri dishes and allow to cure 24 hours at room temperature. Kit includes enough 2-Part Sylgard silicone elastomer to prepare 20 dishes, pins and 20 plastic petri dishes with lids, 65 mm.
Catalog No. NC9666100 Supplier World Precision Instrument Supplier No. 501986
Make your own silicone dissecting pads easily and quickly. Mix the 2-part silicone right in the plastic petri dishes and allow to cure 24 hours at room temperature. Kit includes enough 2-Part Sylgard silicone elastomer to prepare 20 dishes, pins and 20 plastic petri dishes with lids, 65 mm.