Learn More
Med Vet International Silicone Endotracheal Tube w/Connector, 12mm, J0615K
Catalog No. 5000581391
Encompass_Preferred
Endotracheal Tube. 12mm i.d. 51fr 17.0mm o.d. 44cm L balloon 5cm L
Catalog No. 50-005-81391 Supplier Med Vet International Supplier No. JOR615K
Endotracheal Tube. 12mm i.d. 51fr 17.0mm o.d. 44cm L balloon 5cm L