Learn More
Med Vet International Silicone Endotracheal Tube w/Connector, 14mm, J0615M
Catalog No. 5000581445
Encompass_Preferred
Endotracheal Tube. 14mm i.d. 57fr 18.0mm o.d. 48cm L balloon 5cm L
Catalog No. 50-005-81445 Supplier Med Vet International Supplier No. JOR615M
Endotracheal Tube. 14mm i.d. 57fr 18.0mm o.d. 48cm L balloon 5cm L