Learn More
Med Vet International Silicone Endotracheal Tube w/Connector, 4mm, J0615B
Catalog No. 5000581392
Encompass_Preferred
Endotracheal Tube. 4mm i.d. 18fr 6.0mm o.d. 18cm L balloon 3cm L
Catalog No. 50-005-81392 Supplier Med Vet International Supplier No. JOR615B
Endotracheal Tube. 4mm i.d. 18fr 6.0mm o.d. 18cm L balloon 3cm L