Learn More
Med Vet International Silicone Foley Catheter, 22 Fr, Sterile, J0693J
Catalog No. 5000582514
Encompass_Preferred
Clear. 22fr o.d. x 40cm L. 5cc balloon Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation.
Catalog No. 50-005-82514 Supplier Med Vet International Supplier No. JOR693J
Clear. 22fr o.d. x 40cm L. 5cc balloon Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation.