Learn More
Med Vet International Silicone Foley Catheter, 2WAY, w/Stylet, 6FR x 90cm, J0693WL
Catalog No. 5000582195
Encompass_Preferred
Clear. 6fr o.d. x 90cm L. 1.5cc balloon Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation.
Catalog No. 50-005-82195 Supplier Med Vet International Supplier No. JOR693WL
Clear. 6fr o.d. x 90cm L. 1.5cc balloon Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation.