Learn More
Med Vet International Silicone Foley Catheter, 2WAY, w/Stylet, 8FR x 90cm, J0693XL
Catalog No. 5000582512
Encompass_Preferred
Clear. 8fr o.d. x 90cm L. 3.0cc balloon Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation.
Catalog No. 50-005-82512 Supplier Med Vet International Supplier No. JOR693XL
Clear. 8fr o.d. x 90cm L. 3.0cc balloon Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation.