Learn More
Med Vet International Silicone Foley Catheter 5fr, Non-Sterile 30cm, J0693
Catalog No. 5000582513
Encompass_Preferred
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.
Catalog No. 50-005-82513 Supplier Med Vet International Supplier No. JOR693
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.