Learn More
Med Vet International Silicone Foley Catheter 5fr x 55cm, 2cc Balloon Clear, J0693W
Catalog No. 5000582120
Encompass_Preferred
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.NOTE: Balloon should be filled with sterile water or saline for best long-term results.
Catalog No. 50-005-82120 Supplier Med Vet International Supplier No. JOR693W
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.NOTE: Balloon should be filled with sterile water or saline for best long-term results.