Learn More
Med Vet International Silicone Foley Catheter 6fr, Sterile, 30cm, J0693A
Catalog No. 5000582164
Encompass_Preferred
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.
Catalog No. 50-005-82164 Supplier Med Vet International Supplier No. JOR693A
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.