Learn More
Med Vet International Silicone Foley Catheter, Sterile, 12fr x 40cm, 5cc Balloon, J0693D
Catalog No. 5000581742
Encompass_Preferred
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.
Catalog No. 50-005-81742 Supplier Med Vet International Supplier No. JOR693D
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.