Learn More
Med Vet International Silicone Foley Catheter, Sterile, 8fr x 30cm, 3cc Balloon, J0693B
Catalog No. 5000582515
Encompass_Preferred
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.
Catalog No. 50-005-82515 Supplier Med Vet International Supplier No. JOR693B
Silicone is the preferred material for foley catheters. This catheter softens at body temperature. Equipped with a one-way valve for balloon inflation. Sterile.