Learn More
BINDER™ Door Gasket
SDP
Catalog No. 08111650 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers FD 23
Catalog No. For Use With (Equipment)
08-111-650 BINDER Chambers FD 23
1 options
Catalog No. 08-111-650 Supplier BINDER™ Supplier No. 80121306
Add to Cart
Add to Cart
Material: FKM Fluorocarbon; For Use With: FD 23
  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications
Description Door gasket - made of FKM, silicone-free
For Use With (Equipment) BINDER Chambers FD 23
Material FKM Fluorocarbon
Type Door Gasket