Learn More
BINDER™ Door Gasket
SDP
Catalog No. 08111663 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
FKM Fluorocarbon
Catalog No. Material
08-111-663 FKM Fluorocarbon
1 options
Catalog No. 08-111-663 Supplier BINDER™ Supplier No. 80121652
Add to Cart
Add to Cart
Material: FKM Fluorocarbon; For Use With: BINDER units FD 260
  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications
Description Door gasket - made of FKM, silicone-free
For Use With (Equipment) BINDER Chambers FD 260
Material FKM Fluorocarbon
Type Door Gasket