Learn More
BINDER™ Door Gasket
SDP
Catalog No. 08111641 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
FKM Fluorocarbon
Catalog No. Material
08-111-641 FKM Fluorocarbon
1 options
Catalog No. 08-111-641 Supplier BINDER™ Supplier No. 80120493
Add to Cart
Add to Cart
Material: FKM Fluorocarbon; For Use With: BINDER units ED 23
  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications
Description Door gasket - made of FKM, silicone-free
For Use With (Equipment) BINDER Chambers ED 23
Material FKM Fluorocarbon
Type Door Gasket