Learn More
BINDER™ Door Gasket
SDP
Catalog No. 08111645 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
FKM Fluorocarbon
Catalog No. Material
08-111-645 FKM Fluorocarbon
1 options
Catalog No. 08-111-645 Supplier BINDER™ Supplier No. 80120497
Add to Cart
Add to Cart
Material: FKM Fluorocarbon; For Use With: BINDER units ED 400
  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications
Description Door gasket - made of FKM, silicone-free
For Use With (Equipment) BINDER Chambers ED 400
Material FKM Fluorocarbon
Type Door Gasket