Learn More
BINDER™ Door Gasket
SDP
Catalog No. 08111647 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers ED 56
Catalog No. For Use With (Equipment)
08-111-647 BINDER Chambers ED 56
1 options
Catalog No. 08-111-647 Supplier BINDER™ Supplier No. 80121008
Add to Cart
Add to Cart
Material: FKM Fluorocarbon; For Use With: BINDER units ED 56
  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications
Description Door gasket - made of FKM, silicone-free
For Use With (Equipment) BINDER Chambers ED 56
Material FKM Fluorocarbon
Temperature Range (Metric) Ambient to 200°C
Type Door Gasket