Learn More
Brookfield Engineer Labs General Purpose Silicone Fluids, 30000 CPS
Catalog No. NC1530256
Encompass
Brookfield Viscosity Standards provide a convenient, reliable way to verify the calibration of your Brookfield Laboratory Viscometer/Rheometer. The fluids are stored in glass bottles. Silicone viscosity standards supplied in ½ liter (1 pint) glass jars complete with a certificate of calibration.
Catalog No. NC1530256 Supplier Brookfield Engineer Labs Supplier No. 30000 CPS
Brookfield Viscosity Standards provide a convenient, reliable way to verify the calibration of your Brookfield Laboratory Viscometer/Rheometer. The fluids are stored in glass bottles. Silicone viscosity standards supplied in ½ liter (1 pint) glass jars complete with a certificate of calibration.