Learn More
Bellco Glass, Inc. Silicone Headplate O-Ring,10L #274
SDP
Catalog No. 501944908
Encompass_Preferred
Silicone headplate o-ring for 10l bioreactor systems. MADE IN THE USA. Material: red silicone rubber Dimensions: 9.984" ID 10.262" OD Size: #274
Catalog No. 50-194-4908 Supplier Bellco Glass, Inc. Supplier No. 196488810
Silicone headplate o-ring for 10l bioreactor systems. MADE IN THE USA. Material: red silicone rubber Dimensions: 9.984" ID 10.262" OD Size: #274