Learn More
Bellco Glass, Inc. Silicone Headplate O-Ring,3L #254
SDP
Catalog No. 501944906
Encompass_Preferred
Silicone headplate o-ring for 3l bioreactor systems. MADE IN THE USA. Size: 254 Material: red silicone rubber Dimensions: od x id: 5.762"x 0.139" thick
Catalog No. 50-194-4906 Supplier Bellco Glass, Inc. Supplier No. 196488803
Silicone headplate o-ring for 3l bioreactor systems. MADE IN THE USA. Size: 254 Material: red silicone rubber Dimensions: od x id: 5.762"x 0.139" thick