Learn More
Bellco Glass, Inc. Silicone Headplate O-Ring,7L
SDP
Catalog No. 501944907
Encompass_Preferred
Silicone headplate o-ring for 7l bioreactor systems. MADE IN THE USA. Material: red silicone rubber Dimensions: 7.984" ID 8.262" OD Size: #266
Catalog No. 50-194-4907 Supplier Bellco Glass, Inc. Supplier No. 196488807
Silicone headplate o-ring for 7l bioreactor systems. MADE IN THE USA. Material: red silicone rubber Dimensions: 7.984" ID 8.262" OD Size: #266