Learn More
Bellco Glass, Inc. Silicone Heating Pad,7Sq-115V For Bellco Temp Controllers"
SDP
Catalog No. 501944822
Encompass_Preferred
Ideal for use with bellco temperature controllers. MADE IN THE USA. Dimensions: 7" sqMaterial: Red Silicone / Foam backThickness: 3/8"For spinner vessels up to 3L150 Watts115VAC includes 6' power cordWith IEC 60320 C14 male plugHigh limit cutoff up to 225°FConnects to all Bellco Controllers (Temperature controllers)
Catalog No. 50-194-4822 Supplier Bellco Glass, Inc. Supplier No. 196457007
Ideal for use with bellco temperature controllers. MADE IN THE USA. Dimensions: 7" sqMaterial: Red Silicone / Foam backThickness: 3/8"For spinner vessels up to 3L150 Watts115VAC includes 6' power cordWith IEC 60320 C14 male plugHigh limit cutoff up to 225°FConnects to all Bellco Controllers (Temperature controllers)