Learn More
Formulatrix SILICONE INPUT TUBE HOLDER
SDP
Catalog No. NC2311633
Encompass
This tube holder attaches to 15mL or 50mL conical tubes and holds the TEMPEST input lines in place. Input lines can be adjusted to a desired height within the source reservoir and will stay in place during operation.
Catalog No. NC2311633 Supplier Formulatrix Supplier No. 233918B
This tube holder attaches to 15mL or 50mL conical tubes and holds the TEMPEST input lines in place. Input lines can be adjusted to a desired height within the source reservoir and will stay in place during operation.