Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Spooled Precision Pump Tubing, Peroxide-Cured Silicone, L/S 16; 250 ft
Catalog No. NC9811904
Encompass
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
Catalog No. NC9811904 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640716
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.