Learn More
LICORbio Silicone Mats (13cm x 13cm mat)
Catalog No. NC1255518
Encompass_Preferred
These silicone mats are used for flattening membranes against the glass surface on the Odyssey DLx, Odyssey M, Odyssey CLx or Odyssey Classic Infrared Imaging System. The mats come in three sizes: 25 cm x 25 cm, 13 cm x 13 cm, and 19 cm x 26 cm. The 19 cm x 26 cm size is specific for the Odyssey M Imager.
Catalog No. NC1255518 Supplier LICORbio Supplier No. 92670000
These silicone mats are used for flattening membranes against the glass surface on the Odyssey DLx, Odyssey M, Odyssey CLx or Odyssey Classic Infrared Imaging System. The mats come in three sizes: 25 cm x 25 cm, 13 cm x 13 cm, and 19 cm x 26 cm. The 19 cm x 26 cm size is specific for the Odyssey M Imager.