Learn More
World Precision Instrument Silicone Mat, 38x25.5cm
SDP
Catalog No. 50822434
Encompass
  • 38 x 25.5 cm (15 x 10")
  • Autoclavable
  • Instruments sold separately
  • This silicone mat can protect delicate surgical instruments by gently (but firmly) cushioning them and helping to keep them in place to prevent damages.
Catalog No. 50-822-434 Supplier World Precision Instrument Supplier No. 500255
  • 38 x 25.5 cm (15 x 10")
  • Autoclavable
  • Instruments sold separately
  • This silicone mat can protect delicate surgical instruments by gently (but firmly) cushioning them and helping to keep them in place to prevent damages.