Learn More
LICORbio Silicone Mat (19cm x 26cm mat)
Catalog No. NC2225472
Encompass_Preferred
These silicone mats are used for flattening membranes against the glass surface on the Odyssey DLx, Odyssey M, Odyssey CLx or Odyssey Classic Infrared Imaging System. The mats come in three sizes: 25 cm x 25 cm, 13 cm x 13 cm, and 19 cm x 26 cm. The 19 cm x 26 cm size is specific for the Odyssey M Imager.
This item is not returnable. View return policy
Catalog No. NC2225472 Supplier LICORbio Supplier No. 92670003
These silicone mats are used for flattening membranes against the glass surface on the Odyssey DLx, Odyssey M, Odyssey CLx or Odyssey Classic Infrared Imaging System. The mats come in three sizes: 25 cm x 25 cm, 13 cm x 13 cm, and 19 cm x 26 cm. The 19 cm x 26 cm size is specific for the Odyssey M Imager.