Learn More
LICORbio Silicone Mats (25cm x 25cm mat)
Catalog No. NC9304702
Encompass_Preferred
These silicone mats are used for flattening membranes against the glass surface on the Odyssey DLx, Odyssey M, Odyssey CLx or Odyssey Classic Infrared Imaging System. The mats come in three sizes: 25 cm x 25 cm, 13 cm x 13 cm, and 19 cm x 26 cm. The 19 cm x 26 cm size is specific for the Odyssey M Imager.
Catalog No. NC9304702 Supplier LICORbio Supplier No. 92670001
These silicone mats are used for flattening membranes against the glass surface on the Odyssey DLx, Odyssey M, Odyssey CLx or Odyssey Classic Infrared Imaging System. The mats come in three sizes: 25 cm x 25 cm, 13 cm x 13 cm, and 19 cm x 26 cm. The 19 cm x 26 cm size is specific for the Odyssey M Imager.