Learn More
Electron Microscopy Sciences Reusable Silicone Rubber Mold Rectangular, 2.25 x 1"
SDP
Catalog No. 501926838
Encompass_Preferred
These reusable molds for cold embedding are made from sturdy, flexible, white silicone, and are easily removed without the need for release agents. Much smoother than rubber molds, enabling easy identification of samples in clear resins. They hold their shape to produce consistent, reliable round embedding.
Size: 2.25 x 1"
Catalog No. 50-192-6838 Supplier Electron Microscopy Sciences Supplier No. 5048417
These reusable molds for cold embedding are made from sturdy, flexible, white silicone, and are easily removed without the need for release agents. Much smoother than rubber molds, enabling easy identification of samples in clear resins. They hold their shape to produce consistent, reliable round embedding.
Size: 2.25 x 1"