Learn More
Electron Microscopy Sciences Reusable Silicone Rubber Mold Rectangular, 3 x 2"
SDP
Catalog No. 501926839
Encompass_Preferred
These reusable molds for cold embedding are made from sturdy, flexible, white silicone, and are easily removed without the need for release agents. Much smoother than rubber molds, enabling easy identification of samples in clear resins. They hold their shape to produce consistent, reliable round embedding.
Size: 3 x 2"
Catalog No. 50-192-6839 Supplier Electron Microscopy Sciences Supplier No. 5048418
These reusable molds for cold embedding are made from sturdy, flexible, white silicone, and are easily removed without the need for release agents. Much smoother than rubber molds, enabling easy identification of samples in clear resins. They hold their shape to produce consistent, reliable round embedding.
Size: 3 x 2"