Learn More
Electron Microscopy Sciences Reusable Silicone Rubber Mold Round, 1" diameter
SDP
Catalog No. 501926834
Encompass_Preferred
These reusable molds for cold embedding are made from sturdy, flexible, white silicone, and are easily removed without the need for release agents. Much smoother than rubber molds, enabling easy identification of samples in clear resins. They hold their shape to produce consistent, reliable round embedding.
Size: 1" diameter
Catalog No. 50-192-6834 Supplier Electron Microscopy Sciences Supplier No. 5048410
These reusable molds for cold embedding are made from sturdy, flexible, white silicone, and are easily removed without the need for release agents. Much smoother than rubber molds, enabling easy identification of samples in clear resins. They hold their shape to produce consistent, reliable round embedding.
Size: 1" diameter