Learn More
Med Vet International Silicone Nasogastric Feeding Tube, J0386G, 8Fr, 2.7mm x 120cm, Each
Catalog No. 5000582698
Encompass_Preferred
These silicone feeding tubes are great for administering medication and other fluids orally or nasally. All catheters are fully autoclavable. Lateral eyes are on distal tip. Hinged-attached cap.
Catalog No. 50-005-82698 Supplier Med Vet International Supplier No. JOR386G
These silicone feeding tubes are great for administering medication and other fluids orally or nasally. All catheters are fully autoclavable. Lateral eyes are on distal tip. Hinged-attached cap.