Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Silicone O-Ring Seal
SDP

Catalog No. 13900226 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Sealing Jacket Trap into G5 Condenser
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-900-226 Sealing Jacket Trap into G5 Condenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-900-226 Supplier Heidolph™ Supplier No. 036210700
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heidolph™ Silicone O-Ring Seal for use with G5 Glassware

  • Seals jacket trap into G5 condenser

Specifications

Color Red
Material Silicone
Type O-ring
For Use With (Equipment) Sealing Jacket Trap into G5 Condenser
Shape Circle
Dimensions (L x W) 110 x 4 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.