Learn More
Analytical Sales & Services Silicone O-rings for 48-Well TCR
Catalog No. 5000807209
Encompass
Silicone O-rings for 48-Well TCR\n\nOutside diameter: 9mm\nInside diameter: 6mm\nThickness: 1.5mm\n\nUnits/Pkg: 50pk
Catalog No. 50-008-07209 Supplier Analytical Sales & Services Supplier No. 489908
Silicone O-rings for 48-Well TCR\n\nOutside diameter: 9mm\nInside diameter: 6mm\nThickness: 1.5mm\n\nUnits/Pkg: 50pk