Learn More
Lovibond™ Silicone Oil
Catalog No. 09054120 Shop All Tintometer Group Products
Change view
Click to view available options
For Use With (Equipment):
Turbidimeter
Catalog No. For Use With (Equipment)
09-054-120 Turbidimeter
1 options
This item is not returnable. View return policy
Catalog No. 09-054-120 Supplier Lovibond™ Supplier No. 194295
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Silicone Oil is for glassware preparation of Lovibond™ TB Series turbidimeters.
  • Silicone oil can be used to prepare sample cells for turbidity measurement.
  • The oil can be used to mask small scratches and other imperfections in the glassware.
Specifications
Type Silicone Oil
For Use With (Equipment) Turbidimeter
For Use With (Application) Preparation of sample vial for turbidity measurement.
Recommended Accessories Lovibond cleaning cloth (197635)
Volume (English) 15 mL
Weight (English) <1 lb.
Quantity 1 Ea.
Packaging Type LDPE Dropper Bottle