Learn More
Hach Company Silicone Oil, 15 mL SCDB
Supplier:  Hach Company 126936
Encompass
Multiple uses, including preparation of turbidity sample cells, for TL23, TU52, 2100Q, 2100N, 2100AN series
Catalog No. 50-199-4549