Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicone Oil

Catalog No. 175633500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-563-3500G 500 g
17-563-325KG 2.5 kg
17-563-31KG 1 kg
17-563-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-563-3500G Supplier Sigma Aldrich Supplier No. 175633500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicone oil is a highly viscous oil made up of poly(dimethylsiloxane-co-methylphenylsiloxane) chains.Silicone oil has been used as a medium component in containerless protein crystallization techniques which do not require contact with a solid surface. It was used as an oil bath during the synthesis of methyl 2-methylazulene-1-carboxylate.

Specifications

CAS 63148-52-7
Density 1.05 g/mL at 25°C
Refractive Index n20/D 1.495
Linear Formula [Si(CH3)2O]x[Si(CH3)(C6H5)O]y
MDL Number MFCD00133270
Quantity 500 g
Synonym Poly(dimethylsiloxane-co-methylphenylsiloxane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.