Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Silicone oil is low viscosity liquid polymerized siloxanes with organic side chains.Low viscosity silicone oil has been used to prepare cellulose based electrorheological particles for medical devices. The diffusive behavior of spreading of a silicone oil polymer on solid surfaces was studied. It was also used to investigate Marangoni flow, induced by two liquid droplets on a solid surface, exposed in air. Many diverse applications of silicone oil are as heat exchangers, hydrophobic coating of glass, ceramics, textiles and leathers, component of salves.
Specifications
Specifications
| CAS | 63148-62-9 |
| Density | 0.913 g/mL (at 25°C) |
| Boiling Point | >140°C (0.002 mmHg) |
| Linear Formula | [-Si(CH3)2 O-]n |
| Refractive Index | n20/D 1.403 (literature) |
| MDL Number | MFCD00132673 |
| Quantity | 5 mL |
| Synonym | PDMS |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?